missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Thiocyanate 98.0+%, TCI America™

Catalog No. p-7136140
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T020025ML 25 mL
T0200500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T020025ML Supplier TCI America Supplier No. T020025ML

Chemical Identifiers

CAS 542-90-5
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00001834
InChI Key WFCLYEAZTHWNEH-UHFFFAOYSA-N
Synonym ethylthiocyanate, thiocyanic acid, ethyl ester, ethyl rhodanate, ethyl sulfocyanate, aethylrhodanid, ethane, thiocyanato, ethylthiokyanat, aethylrhodanid german, ethylthiokyanat czech, ethyl rhodanide
PubChem CID 10968
IUPAC Name ethyl thiocyanate
SMILES CCSC#N

Specifications

Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 1992
Formula Weight 87.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Thiocyanate
RTECSNumber XK9900000
TSCA Yes