Learn More
Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%
CAS: 312745-91-8 | C7H11NO3 | 157.17 g/mol
Supplier: Thermo Scientific Chemicals L2025914
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible or difficult to mix with water.
Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate | |
| 1.114 | |
| >110°C (230°F) | |
| C7H11NO3 | |
| MFCD00672878 | |
| ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112 | |
| LOQFROBMBSKWQY-UHFFFAOYNA-N | |
| CCOC(=O)CC(O)CC#N | |
| 157.17 | |
| 157.17 |
| 312745-91-8 | |
| 270°C | |
| ee ≥98% | |
| 1.448 | |
| 25 g | |
| Not miscible or difficult to mix with water. | |
| +5° (Neat) | |
| ethyl (3S)-4-cyano-3-hydroxybutanoate | |
| 7020868 | |
| 97% |
Chemical Identifiers
| 312745-91-8 | |
| 157.17 | |
| LOQFROBMBSKWQY-UHFFFAOYNA-N | |
| 7020868 | |
| CCOC(=O)CC(O)CC#N |
| C7H11NO3 | |
| MFCD00672878 | |
| ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112 | |
| ethyl (3S)-4-cyano-3-hydroxybutanoate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only