missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%

Catalog No. p-7048122
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2025906 5 g
AAL2025914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2025906 Supplier Thermo Scientific Chemicals Supplier No. L2025906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 312745-91-8 | C7H11NO3 | 157.17 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 312745-91-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00672878
InChI Key LOQFROBMBSKWQY-UHFFFAOYNA-N
Synonym ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112
PubChem CID 7020868
IUPAC Name ethyl (3S)-4-cyano-3-hydroxybutanoate
SMILES CCOC(=O)CC(O)CC#N

Specifications

Density 1.114
Boiling Point 270°C
Flash Point >110°C (230°F)
Refractive Index 1.448
Quantity 5 g
Solubility Information Not miscible or difficult to mix with water.
Optical Rotation +5° (Neat)
Formula Weight 157.17
Percent Purity 97%
Assay ee ≥98%
Chemical Name or Material Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only