accessibility menu, dialog, popup

UserName

CAS RN 312745-91-8

IUPAC Name: ethyl 4-cyano-3-hydroxybutanoate
Synonyms: ethyl 4-cyano-3-hydroxybutanoate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: LOQFROBMBSKWQY-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(O)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results