missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methyl Sulfide 98.0+%, TCI America™
$59.69 - $154.02
Chemical Identifiers
| CAS | 624-89-5 |
|---|---|
| Molecular Formula | C3H8S |
| Molecular Weight (g/mol) | 76.16 |
| MDL Number | MFCD00009268 |
| InChI Key | WXEHBUMAEPOYKP-UHFFFAOYSA-N |
| Synonym | ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane |
| PubChem CID | 12230 |
| IUPAC Name | (methylsulfanyl)ethane |
| SMILES | CCSC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E014325ML
|
TCI America
E014325ML |
25 mL |
Each for $59.69
|
|
|||||
|
E0143100ML
|
TCI America
E0143100ML |
100 mL |
Each for $154.02
|
|
|||||
Chemical Identifiers
| 624-89-5 | |
| 76.16 | |
| WXEHBUMAEPOYKP-UHFFFAOYSA-N | |
| 12230 | |
| CCSC |
| C3H8S | |
| MFCD00009268 | |
| ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane | |
| (methylsulfanyl)ethane |
Specifications
| 624-89-5 | |
| 66°C | |
| MFCD00009268 | |
| 1993 | |
| WXEHBUMAEPOYKP-UHFFFAOYSA-N | |
| (methylsulfanyl)ethane | |
| 12230 | |
| ≥98.0% (GC) | |
| Ethyl Methyl Sulfide |
| Yellow | |
| C3H8S | |
| 25 mL | |
| ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane | |
| CCSC | |
| 76.16 | |
| 76.16 | |
| Liquid |
Safety and Handling
TSCA : Yes