missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methyl Sulfide 98.0+%, TCI America™
Supplier: TCI America E0143100ML
Specifications
| Ethyl Methyl Sulfide | |
| Yellow | |
| C3H8S | |
| 100 mL | |
| ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane | |
| CCSC | |
| 76.16 | |
| 76.16 | |
| Liquid |
| 624-89-5 | |
| 66°C | |
| MFCD00009268 | |
| 1993 | |
| WXEHBUMAEPOYKP-UHFFFAOYSA-N | |
| (methylsulfanyl)ethane | |
| 12230 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 624-89-5 | |
| 76.16 | |
| WXEHBUMAEPOYKP-UHFFFAOYSA-N | |
| 12230 | |
| CCSC |
| C3H8S | |
| MFCD00009268 | |
| ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane | |
| (methylsulfanyl)ethane |
Safety and Handling
TSCA : Yes