missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methanesulfonate 99.0+%, TCI America™
$95.06 - $236.73
Chemical Identifiers
| CAS | 62-50-0 |
|---|---|
| Molecular Formula | C3H8O3S |
| Molecular Weight (g/mol) | 124.154 |
| MDL Number | MFCD00007559 |
| InChI Key | PLUBXMRUUVWRLT-UHFFFAOYSA-N |
| Synonym | ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid |
| PubChem CID | 6113 |
| ChEBI | CHEBI:23994 |
| IUPAC Name | ethyl methanesulfonate |
| SMILES | CCOS(=O)(=O)C |
Chemical Identifiers
| 62-50-0 | |
| 124.154 | |
| PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
| 6113 | |
| ethyl methanesulfonate |
| C3H8O3S | |
| MFCD00007559 | |
| ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
| CHEBI:23994 | |
| CCOS(=O)(=O)C |
Specifications
| 62-50-0 | |
| 89°C | |
| MFCD00007559 | |
| ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
| CCOS(=O)(=O)C | |
| 124.154 | |
| CHEBI:23994 | |
| ≥99.0% (GC) | |
| Ethyl Methanesulfonate |
| Yellow | |
| C3H8O3S | |
| 25 g | |
| PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
| ethyl methanesulfonate | |
| 6113 | |
| 124.15 | |
| Liquid |
Safety and Handling
RTECSNumber : PB2100000
TSCA : Yes