accessibility menu, dialog, popup

UserName

CH3OSCH3OO

CAS RN 62-50-0

IUPAC Name: ethyl methanesulfonate
Synonyms: ethyl methanesulfonate
Molecular Weight (g/mol): 124.15
Molecular Formula: C3H8O3S
InChi Key: PLUBXMRUUVWRLT-UHFFFAOYSA-N
SMILES: CCOS(C)(=O)=O
Boiling Point 
  • (4)
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (8)
  • (4)
  • (1)
  • (3)
  • (2)
  • (4)
  • (6)
  • (2)
  • (8)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)