missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methanesulfonate 99.0+%, TCI America™
Supplier: TCI America M0607100G
Specifications
| Ethyl Methanesulfonate | |
| Yellow | |
| C3H8O3S | |
| 100 g | |
| PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
| ethyl methanesulfonate | |
| 6113 | |
| 124.15 | |
| Liquid |
| 62-50-0 | |
| 89°C | |
| MFCD00007559 | |
| ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
| CCOS(=O)(=O)C | |
| 124.154 | |
| CHEBI:23994 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 62-50-0 | |
| 124.154 | |
| PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
| 6113 | |
| ethyl methanesulfonate |
| C3H8O3S | |
| MFCD00007559 | |
| ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
| CHEBI:23994 | |
| CCOS(=O)(=O)C |
Safety and Handling
RTECSNumber : PB2100000
TSCA : Yes