missing translation for 'onlineSavingsMsg'
Learn More

Ethyl mesitylglyoxylate, 97%

Catalog No. p-7058392
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1310606 5 g
AAL1310614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1310606 Supplier Thermo Scientific Chemicals Supplier No. L1310606
Only null left

CAS: 5524-57-2 | C13H16O3 | 220.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5524-57-2
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
MDL Number MFCD00151821
InChI Key XRSPPFOBPRPYLD-UHFFFAOYSA-N
Synonym ethyl mesitylglyoxylate, ethyl 2-mesityl-2-oxoacetate, ethyl 2-oxo-2-2,4,6-trimethylphenyl acetate, acmc-1asq7, mesitylglyoxylic acid ethyl ester, ethyl 2,4,6-trimethylphenyl glyoxylate, alpha-oxo-2,4,6-trimethylbenzeneacetic acid ethyl ester
PubChem CID 2736481
IUPAC Name ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
SMILES CCOC(=O)C(=O)C1=C(C=C(C=C1C)C)C

Specifications

Boiling Point 164°C to 165°C (10 mmHg)
Refractive Index 1.512
Quantity 5 g
Beilstein 2970031
Sensitivity Moisture sensitive
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material Ethyl mesitylglyoxylate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only