CAS RN 5524-57-2
IUPAC Name:
ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
Synonyms:
ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
Molecular Weight (g/mol):
220.27
Molecular Formula:
C13H16O3
InChi Key:
XRSPPFOBPRPYLD-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)C1=C(C)C=C(C)C=C1C
1
–
1
of
1
results
1
Promotions Available
Ethyl mesitylglyoxylate, 97%
Ethyl mesitylglyoxylate, 97%
CAS: 5524-57-2 Molecular Formula: C13H16O3 Molecular Weight (g/mol): 220.268 MDL Number: MFCD00151821 InChI Key: XRSPPFOBPRPYLD-UHFFFAOYSA-N Synonym: ethyl mesitylglyoxylate,ethyl 2-mesityl-2-oxoacetate,ethyl 2-oxo-2-2,4,6-trimethylphenyl acetate,acmc-1asq7,mesitylglyoxylic acid ethyl ester,ethyl 2,4,6-trimethylphenyl glyoxylate,alpha-oxo-2,4,6-trimethylbenzeneacetic acid ethyl ester PubChem CID: 2736481 IUPAC Name: ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate SMILES: CCOC(=O)C(=O)C1=C(C=C(C=C1C)C)C