missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl isothiocyanate, 97%
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Reacts with water. Soluble in chlorinated and aromatic hydrocarbons. Miscible in ethanol and ether.
Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is used as a pharmaceutical intermediate.
Solubility
Reacts with water. Soluble in chlorinated and aromatic hydrocarbons. Miscible in ethanol and ether.
Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Melting Point | -6°C |
| Density | 0.995 |
| Boiling Point | 130°C to 132°C |
| Flash Point | 32°C (89°F) |
| Odor | Pungent |
| Refractive Index | 1.511 |
| Quantity | 25 g |
| UN Number | UN2929 |
| Beilstein | 1737705 |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H226-H301+H311+H331-H314-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H300-H310-H330-H334-H314-H318-H226 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| EINECSNumber | 208-831-2 |
| RTECSNumber | NX8590000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only