missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isothiocyanate 97.0+%, TCI America™

$93.21 - $93.21

Chemical Identifiers

CAS 542-85-8
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00004820
InChI Key HBNYJWAFDZLWRS-UHFFFAOYSA-N
Synonym ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
PubChem CID 10966
ChEBI CHEBI:85098
IUPAC Name isothiocyanatoethane
SMILES CCN=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I018825ML
View Documents
TCI America
I018825ML
25 mL
Each for $93.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

542-85-8
87.14
HBNYJWAFDZLWRS-UHFFFAOYSA-N
10966
isothiocyanatoethane
C3H5NS
MFCD00004820
ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
CHEBI:85098
CCN=C=S

Specifications

542-85-8
132°C
MFCD00004820
1992
HBNYJWAFDZLWRS-UHFFFAOYSA-N
isothiocyanatoethane
10966
87.14
Liquid
Yellow
C3H5NS
25 mL
ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
CCN=C=S
87.14
CHEBI:85098
≥97.0% (GC)
Ethyl Isothiocyanate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NX8590000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'