missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Isopropyl Sulfone 97.0+%, TCI America™
$72.70 - $275.87
Chemical Identifiers
| CAS | 4853-75-2 |
|---|---|
| Molecular Formula | C5H12O2S |
| Molecular Weight (g/mol) | 136.209 |
| MDL Number | MFCD00050418 |
| InChI Key | RDKKQZIFDSEMNU-UHFFFAOYSA-N |
| PubChem CID | 21719919 |
| IUPAC Name | 2-ethylsulfonylpropane |
| SMILES | CCS(=O)(=O)C(C)C |
Chemical Identifiers
| 4853-75-2 | |
| 136.209 | |
| RDKKQZIFDSEMNU-UHFFFAOYSA-N | |
| 2-ethylsulfonylpropane |
| C5H12O2S | |
| MFCD00050418 | |
| 21719919 | |
| CCS(=O)(=O)C(C)C |
Specifications
| 4853-75-2 | |
| 91°C | |
| MFCD00050418 | |
| RDKKQZIFDSEMNU-UHFFFAOYSA-N | |
| 2-ethylsulfonylpropane | |
| 21719919 | |
| ≥97.0% (GC) | |
| Ethyl Isopropyl Sulfone |
| Yellow | |
| C5H12O2S | |
| 5 g | |
| CCS(=O)(=O)C(C)C | |
| 136.209 | |
| 136.21 | |
| Liquid |
Safety and Handling
TSCA : No