accessibility menu, dialog, popup

UserName

CAS RN 4853-75-2

H3CSOOCH3H3C

CAS RN 4853-75-2

IUPAC Name: 2-(ethanesulfonyl)propane
Synonyms: 2-(ethanesulfonyl)propane
Molecular Weight (g/mol): 136.21
Molecular Formula: C5H12O2S
InChi Key: RDKKQZIFDSEMNU-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results