missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Isopropyl Sulfone 97.0+%, TCI America™
Supplier: TCI America E047525G
Specifications
| Ethyl Isopropyl Sulfone | |
| Yellow | |
| C5H12O2S | |
| 25 g | |
| CCS(=O)(=O)C(C)C | |
| 136.209 | |
| 136.21 | |
| Liquid |
| 4853-75-2 | |
| 91°C | |
| MFCD00050418 | |
| RDKKQZIFDSEMNU-UHFFFAOYSA-N | |
| 2-ethylsulfonylpropane | |
| 21719919 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 4853-75-2 | |
| 136.209 | |
| RDKKQZIFDSEMNU-UHFFFAOYSA-N | |
| 2-ethylsulfonylpropane |
| C5H12O2S | |
| MFCD00050418 | |
| 21719919 | |
| CCS(=O)(=O)C(C)C |
Safety and Handling
TSCA : No