Learn More
Ethyl isocyanatoacetate, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 2949-22-6 |
|---|---|
| Molecular Formula | C5H7NO3 |
| Molecular Weight (g/mol) | 129.12 |
| MDL Number | MFCD00002040 |
| InChI Key | DUVOZUPPHBRJJO-UHFFFAOYSA-N |
| Synonym | ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester |
| PubChem CID | 76283 |
| IUPAC Name | ethyl 2-isocyanatoacetate |
| SMILES | CCOC(=O)CN=C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC410150050
|
Thermo Scientific Chemicals
410150050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC410150250
|
Thermo Scientific Chemicals
410150250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC410151000
|
Thermo Scientific Chemicals
410151000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2949-22-6 | |
| 129.12 | |
| DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
| 76283 | |
| CCOC(=O)CN=C=O |
| C5H7NO3 | |
| MFCD00002040 | |
| ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
| ethyl 2-isocyanatoacetate |
Specifications
| 2949-22-6 | |
| 67°C to 68°C (11.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4210 to 1.4230 (20°C, 589nm) | |
| MFCD00002040 | |
| 1.151 | |
| Solubility in water: hydrolyses | |
| CCOC(=O)CN=C=O | |
| 129.12 | |
| 129.11 | |
| Ethyl isocyanatoacetate |
| 1.1510g/mL | |
| 72°C | |
| 97.5% min. (GC) | |
| C5H7NO3 | |
| OCNCH2COOC2H5 | |
| 5g | |
| ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
| DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
| ethyl 2-isocyanatoacetate | |
| 76283 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause re
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Danger
EINECSNumber : 220-967-4
RUO â Research Use Only