missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isocyanatoacetate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 410150050

 View more versions of this product

Catalog No. AC410150050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl isocyanatoacetate
1.1510g/mL
72°C
97.5% min. (GC)
C5H7NO3
OCNCH2COOC2H5
5g
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
DUVOZUPPHBRJJO-UHFFFAOYSA-N
ethyl 2-isocyanatoacetate
76283
98%
2949-22-6
67°C to 68°C (11.0mmHg)
Authentic
Glass bottle
1.4210 to 1.4230 (20°C, 589nm)
MFCD00002040
1.151
Solubility in water: hydrolyses
CCOC(=O)CN=C=O
129.12
129.11

Chemical Identifiers

2949-22-6
129.12
DUVOZUPPHBRJJO-UHFFFAOYSA-N
76283
CCOC(=O)CN=C=O
C5H7NO3
MFCD00002040
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
ethyl 2-isocyanatoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause re

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice

GHS Signal Word: Danger

EINECSNumber : 220-967-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only