Learn More
Ethyl isocyanatoacetate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 410150250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Ethyl isocyanatoacetate | |
| 1.1510g/mL | |
| 72°C | |
| 97.5% min. (GC) | |
| C5H7NO3 | |
| OCNCH2COOC2H5 | |
| 25g | |
| ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
| DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
| ethyl 2-isocyanatoacetate | |
| 76283 | |
| 98% |
| 2949-22-6 | |
| 67°C to 68°C (11.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4210 to 1.4230 (20°C, 589nm) | |
| MFCD00002040 | |
| 1.151 | |
| Solubility in water: hydrolyses | |
| CCOC(=O)CN=C=O | |
| 129.12 | |
| 129.11 |
Chemical Identifiers
| 2949-22-6 | |
| 129.12 | |
| DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
| 76283 | |
| CCOC(=O)CN=C=O |
| C5H7NO3 | |
| MFCD00002040 | |
| ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
| ethyl 2-isocyanatoacetate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause re
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Danger
EINECSNumber : 220-967-4
RUO â Research Use Only