Learn More
Ethyl isobutyrylacetate, 94%
CAS: 7152-15-0 | C8H14O3 | 158.197 g/mol
Supplier: Thermo Scientific Chemicals L0901614
Description
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.
Solubility
Soluble in chloroform and methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| Ethyl isobutyrylacetate | |
| -9°C | |
| 172°C to 174°C | |
| C8H14O3 | |
| MFCD00009198 | |
| UN3272 | |
| ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate | |
| XCLDSQRVMMXWMS-UHFFFAOYSA-N | |
| ethyl 4-methyl-3-oxopentanoate | |
| 81583 | |
| 94% |
| 7152-15-0 | |
| 0.98 | |
| 53°C (127°F) | |
| 1.4265 | |
| 25 g | |
| 636726 | |
| Soluble in chloroform and methanol. | |
| CCOC(=O)CC(=O)C(C)C | |
| 158.197 | |
| 158.2 |
Chemical Identifiers
| 7152-15-0 | |
| 158.197 | |
| XCLDSQRVMMXWMS-UHFFFAOYSA-N | |
| 81583 | |
| CCOC(=O)CC(=O)C(C)C |
| C8H14O3 | |
| MFCD00009198 | |
| ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate | |
| ethyl 4-methyl-3-oxopentanoate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 230-491-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only