accessibility menu, dialog, popup

UserName

H3COOOCH3CH3

CAS RN 7152-15-0

IUPAC Name: ethyl 4-methyl-3-oxopentanoate
Synonyms: ethyl 4-methyl-3-oxopentanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: XCLDSQRVMMXWMS-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)