missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrylacetate, 94%

Catalog No. p-7044991
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0901606 5 g
AAL0901614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0901606 Supplier Thermo Scientific Chemicals Supplier No. L0901606
Only null left

CAS: 7152-15-0 | C8H14O3 | 158.197 g/mol

Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl Isobutyrylacetate is used in the synthesis of piperazine derivatives as possible multireceptor atypical antipsychotics, affecting dopamine and serotonin receptor properties . Also used in the synthesis of pyrazinecarboxamide DGAT1 (diacylglycerol acyltransferase 1) inhibitors used in the treatment of obesity.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 7152-15-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00009198
InChI Key XCLDSQRVMMXWMS-UHFFFAOYSA-N
Synonym ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
PubChem CID 81583
IUPAC Name ethyl 4-methyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)C

Specifications

Melting Point -9°C
Density 0.98
Boiling Point 172°C to 174°C
Flash Point 53°C (127°F)
Refractive Index 1.4265
Quantity 5 g
UN Number UN3272
Beilstein 636726
Solubility Information Soluble in chloroform and methanol.
Formula Weight 158.2
Percent Purity 94%
Chemical Name or Material Ethyl isobutyrylacetate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 230-491-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only