missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Fluoroacetate 98.0+%, TCI America™

Supplier:  TCI America F0030100ML

 View more versions of this product

Catalog No. F0030100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Fluoroacetate
Yellow
C4H7FO2
100 mL
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
CCOC(=O)CF
106.096
106.10
Liquid
459-72-3
117°C
MFCD00000450
2929
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl 2-fluoroacetate
9988
≥98.0% (GC)

Chemical Identifiers

459-72-3
106.096
VCYZVXRKYPKDQB-UHFFFAOYSA-N
9988
CCOC(=O)CF
C4H7FO2
MFCD00000450
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
ethyl 2-fluoroacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3837

RTECSNumber : AH7175000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions