missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Fluoroacetate 98.0+%, TCI America™

Catalog No. p-7129449
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F003025ML 25 mL
F0030100ML 100 mL
F0030500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F003025ML Supplier TCI America Supplier No. F003025ML
Only null left

Chemical Identifiers

CAS 459-72-3
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.096
MDL Number MFCD00000450
InChI Key VCYZVXRKYPKDQB-UHFFFAOYSA-N
Synonym ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
PubChem CID 9988
IUPAC Name ethyl 2-fluoroacetate
SMILES CCOC(=O)CF

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 mL
UN Number 2929
Formula Weight 106.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Fluoroacetate
EINECSNumber (2)-3837
RTECSNumber AH7175000
TSCA Yes