missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Diphenylphosphonoacetate 96.0+%, TCI America™

Catalog No. D25471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25471G 1 g
D25475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25471G Supplier TCI America Supplier No. D25471G

Chemical Identifiers

CAS 16139-79-0
Molecular Formula C16H17O5P
Molecular Weight (g/mol) 320.281
MDL Number MFCD01321165
InChI Key UQMFCYBSUVRGNU-UHFFFAOYSA-N
Synonym Diphenylphosphonoacetic Acid Ethyl Ester
PubChem CID 11056297
IUPAC Name ethyl 2-diphenoxyphosphorylacetate
SMILES CCOC(=O)CP(=O)(OC1=CC=CC=C1)OC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 220°C
Quantity 1 g
Formula Weight 320.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Diphenylphosphonoacetate [Horner-Emmons Reagent]
TSCA No
Recommended Storage Refrigerator