accessibility menu, dialog, popup

UserName

H3COOPOOO

CAS RN 16139-79-0

IUPAC Name: ethyl 2-(diphenoxyphosphoryl)acetate
Synonyms: ethyl 2-(diphenoxyphosphoryl)acetate
Molecular Weight (g/mol): 320.28
Molecular Formula: C16H17O5P
InChi Key: UQMFCYBSUVRGNU-UHFFFAOYSA-N
SMILES: CCOC(=O)CP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results