Learn More
Ethyl cyclohexylideneacetate, 98%
CAS: 1552-92-7 | C10H16O2 | 168.24 g/mol
Supplier: Thermo Scientific Chemicals B2507314
Description
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl cyclohexylideneacetate is used as pharmaceutical intermediates.
Solubility
Soluble in dichloromethane, ethyl acetate.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| Ethyl cyclohexylideneacetate | |
| 48°C to 49°C (0.02 mmHg) | |
| MFCD00017627,MFCD00963857 | |
| ethyl cyclohexylideneacetate, acetic acid, cyclohexylidene-, ethyl ester, ethyl 2-cyclohexylidenacetate, cyclohexylideneacetic acid ethyl ester, ethoxycarbonylmethylenecyclohexane, .delta.1,.alpha.-cyclohexaneacetic acid, ethyl ester, acmc-20ahxr, ethylcyclohexylidenacetate, .delta.1, ethyl ester, ethyl2-cyclohexylideneacetate | |
| MCWDXHYYYNGYGK-UHFFFAOYSA-N | |
| ethyl 2-cyclohexylideneacetate | |
| 73776 | |
| 98% |
| 1552-92-7 | |
| C10H16O2 | |
| 25 g | |
| Soluble in dichloromethane,ethyl acetate. | |
| CCOC(=O)C=C1CCCCC1 | |
| 168.24 | |
| 168.24 |
Chemical Identifiers
| 1552-92-7 | |
| 168.24 | |
| MCWDXHYYYNGYGK-UHFFFAOYSA-N | |
| 73776 | |
| CCOC(=O)C=C1CCCCC1 |
| C10H16O2 | |
| MFCD00017627,MFCD00963857 | |
| ethyl cyclohexylideneacetate, acetic acid, cyclohexylidene-, ethyl ester, ethyl 2-cyclohexylidenacetate, cyclohexylideneacetic acid ethyl ester, ethoxycarbonylmethylenecyclohexane, .delta.1,.alpha.-cyclohexaneacetic acid, ethyl ester, acmc-20ahxr, ethylcyclohexylidenacetate, .delta.1, ethyl ester, ethyl2-cyclohexylideneacetate | |
| ethyl 2-cyclohexylideneacetate |
Safety and Handling
EINECSNumber : 216-297-7
TSCA : No
RUO – Research Use Only