missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyclohexylideneacetate, 98%

Catalog No. p-7027975
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2507306 5 g
AAB2507314 25 g
AAB2507322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2507306 Supplier Thermo Scientific Chemicals Supplier No. B2507306

CAS: 1552-92-7 | C10H16O2 | 168.24 g/mol

Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl acetate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1552-92-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00017627,MFCD00963857
InChI Key MCWDXHYYYNGYGK-UHFFFAOYSA-N
Synonym ethyl cyclohexylideneacetate, acetic acid, cyclohexylidene-, ethyl ester, ethyl 2-cyclohexylidenacetate, cyclohexylideneacetic acid ethyl ester, ethoxycarbonylmethylenecyclohexane, .delta.1,.alpha.-cyclohexaneacetic acid, ethyl ester, acmc-20ahxr, ethylcyclohexylidenacetate, .delta.1, ethyl ester, ethyl2-cyclohexylideneacetate
PubChem CID 73776
IUPAC Name ethyl 2-cyclohexylideneacetate
SMILES CCOC(=O)C=C1CCCCC1

Specifications

Boiling Point 48°C to 49°C (0.02 mmHg)
Quantity 5 g
Solubility Information Soluble in dichloromethane,ethyl acetate.
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl cyclohexylideneacetate
EINECSNumber 216-297-7
TSCA No

RUO – Research Use Only