missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Cyano(hydroxyimino)acetate 98.0+%, TCI America™
Supplier: TCI America E0847100G
Specifications
| Ethyl Cyano(hydroxyimino)acetate | |
| 133°C | |
| C5H6N2O3 | |
| 100 g | |
| LCFXLZAXGXOXAP-QPJJXVBHSA-N | |
| ethyl (2E)-2-cyano-2-hydroxyiminoacetate | |
| 6400537 | |
| ≥98.0% (GC) |
| 3849-21-6 | |
| White-Yellow | |
| MFCD00002112 | |
| ethyl cyanoglyoxylate-2-oxime, ethyl isonitrosocyanoacetate, ethyl cyano hydroxyimino acetate, ethyl hydroxyimino cyanoacetate, ethyl 2-cyano-2-hydroxyimino acetate, ethyl cyano hydroxyimino formate, ethyl oximinocyanoacetate, oxyma, e-ethyl 2-cyano-2-hydroxyimino acetate, acetic acid, cyano hydroxyimino-, ethyl ester | |
| CCOC(=O)C(=NO)C#N | |
| 142.114 | |
| 142.11 | |
| Crystalline Powder |
Chemical Identifiers
| 3849-21-6 | |
| 142.114 | |
| LCFXLZAXGXOXAP-QPJJXVBHSA-N | |
| 6400537 | |
| CCOC(=O)C(=NO)C#N |
| C5H6N2O3 | |
| MFCD00002112 | |
| ethyl cyanoglyoxylate-2-oxime, ethyl isonitrosocyanoacetate, ethyl cyano hydroxyimino acetate, ethyl hydroxyimino cyanoacetate, ethyl 2-cyano-2-hydroxyimino acetate, ethyl cyano hydroxyimino formate, ethyl oximinocyanoacetate, oxyma, e-ethyl 2-cyano-2-hydroxyimino acetate, acetic acid, cyano hydroxyimino-, ethyl ester | |
| ethyl (2E)-2-cyano-2-hydroxyiminoacetate |
Safety and Handling
TSCA : No