missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyano(hydroxyimino)acetate 98.0+%, TCI America™

Catalog No. p-7129182
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E084725G 25 g
E0847100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E084725G Supplier TCI America Supplier No. E084725G
Only null left

Chemical Identifiers

CAS 3849-21-6
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.114
MDL Number MFCD00002112
InChI Key LCFXLZAXGXOXAP-QPJJXVBHSA-N
Synonym ethyl cyanoglyoxylate-2-oxime, ethyl isonitrosocyanoacetate, ethyl cyano hydroxyimino acetate, ethyl hydroxyimino cyanoacetate, ethyl 2-cyano-2-hydroxyimino acetate, ethyl cyano hydroxyimino formate, ethyl oximinocyanoacetate, oxyma, e-ethyl 2-cyano-2-hydroxyimino acetate, acetic acid, cyano hydroxyimino-, ethyl ester
PubChem CID 6400537
IUPAC Name ethyl (2E)-2-cyano-2-hydroxyiminoacetate
SMILES CCOC(=O)C(=NO)C#N

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 25 g
Formula Weight 142.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Cyano(hydroxyimino)acetate