accessibility menu, dialog, popup

UserName

CAS RN 3849-21-6

IUPAC Name: (E)-(ethyl cyano(hydroxyimino)formate)
Synonyms: (E)-(ethyl cyano(hydroxyimino)formate)
Molecular Weight (g/mol): 142.11
Molecular Formula: C5H6N2O3
InChi Key: LCFXLZAXGXOXAP-QPJJXVBHSA-N
SMILES: CCOC(=O)C(=N\O)\C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)