missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Bromofluoroacetate 98.0+%, TCI America™
$55.84 - $217.54
Chemical Identifiers
| CAS | 401-55-8 |
|---|---|
| Molecular Formula | C4H6BrFO2 |
| Molecular Weight (g/mol) | 184.992 |
| MDL Number | MFCD00042095 |
| InChI Key | ULNDTPIRBQGESN-UHFFFAOYSA-N |
| Synonym | ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t |
| PubChem CID | 2733407 |
| IUPAC Name | ethyl 2-bromo-2-fluoroacetate |
| SMILES | CCOC(=O)C(F)Br |
Chemical Identifiers
| 401-55-8 | |
| 184.992 | |
| ULNDTPIRBQGESN-UHFFFAOYSA-N | |
| 2733407 | |
| CCOC(=O)C(F)Br |
| C4H6BrFO2 | |
| MFCD00042095 | |
| ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t | |
| ethyl 2-bromo-2-fluoroacetate |
Specifications
| 401-55-8 | |
| 68°C | |
| MFCD00042095 | |
| 2810 | |
| ULNDTPIRBQGESN-UHFFFAOYSA-N | |
| ethyl 2-bromo-2-fluoroacetate | |
| 2733407 | |
| ≥98.0% (GC) | |
| Ethyl Bromofluoroacetate |
| Yellow | |
| C4H6BrFO2 | |
| 5 g | |
| ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t | |
| CCOC(=O)C(F)Br | |
| 184.992 | |
| 184.99 | |
| Liquid |
Safety and Handling
TSCA : No