accessibility menu, dialog, popup

UserName

CAS RN 401-55-8

IUPAC Name: ethyl 2-bromo-2-fluoroacetate
Synonyms: ethyl 2-bromo-2-fluoroacetate
Molecular Weight (g/mol): 184.99
Molecular Formula: C4H6BrFO2
InChi Key: ULNDTPIRBQGESN-UHFFFAOYNA-N
SMILES: CCOC(=O)C(F)Br
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)