missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Bromodifluoroacetate 98.0+%, TCI America™
Supplier: TCI America B14635G
Specifications
| Ethyl Bromodifluoroacetate | |
| Yellow | |
| MFCD00042069 | |
| 2924 | |
| IRSJDVYTJUCXRV-UHFFFAOYSA-N | |
| ethyl 2-bromo-2,2-difluoroacetate | |
| 69585 | |
| ≥98.0% (GC) |
| 667-27-6 | |
| C4H5BrF2O2 | |
| 5 g | |
| ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953 | |
| CCOC(=O)C(F)(F)Br | |
| 202.983 | |
| 202.98 | |
| Liquid |
Chemical Identifiers
| 667-27-6 | |
| 202.983 | |
| IRSJDVYTJUCXRV-UHFFFAOYSA-N | |
| 69585 | |
| CCOC(=O)C(F)(F)Br |
| C4H5BrF2O2 | |
| MFCD00042069 | |
| ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953 | |
| ethyl 2-bromo-2,2-difluoroacetate |
Safety and Handling
TSCA : No