missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Bromodifluoroacetate 98.0+%, TCI America™

Catalog No. p-7119065
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14635G 5 g
B146325G 25 g
B1463250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B14635G Supplier TCI America Supplier No. B14635G
Only null left

Chemical Identifiers

CAS 667-27-6
Molecular Formula C4H5BrF2O2
Molecular Weight (g/mol) 202.983
MDL Number MFCD00042069
InChI Key IRSJDVYTJUCXRV-UHFFFAOYSA-N
Synonym ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953
PubChem CID 69585
IUPAC Name ethyl 2-bromo-2,2-difluoroacetate
SMILES CCOC(=O)C(F)(F)Br

Specifications

Color Yellow
Quantity 5 g
UN Number 2924
Formula Weight 202.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Bromodifluoroacetate