missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Benzilate 97.0+%, TCI America™
$635.31 - $1,089.64
Chemical Identifiers
| CAS | 52182-15-7 |
|---|---|
| Molecular Formula | C16H16O3 |
| Molecular Weight (g/mol) | 256.30 |
| MDL Number | MFCD00016850 |
| InChI Key | AIPVNQQMYPWQSX-UHFFFAOYSA-N |
| Synonym | ethyl benzilate, benzilic acid ethyl ester, ethyl diphenylglycolate, ethylbenzilate, ethyl benzillate, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, ethyl ester, benzilsaureethylester, acmc-209ky4, ethyl hydroxy diphenyl acetate #, diphenylglycolic acid ethyl ester |
| PubChem CID | 96261 |
| IUPAC Name | ethyl 2-hydroxy-2,2-diphenylacetate |
| SMILES | CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 52182-15-7 | |
| 256.30 | |
| AIPVNQQMYPWQSX-UHFFFAOYSA-N | |
| 96261 | |
| CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C16H16O3 | |
| MFCD00016850 | |
| ethyl benzilate, benzilic acid ethyl ester, ethyl diphenylglycolate, ethylbenzilate, ethyl benzillate, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, ethyl ester, benzilsaureethylester, acmc-209ky4, ethyl hydroxy diphenyl acetate #, diphenylglycolic acid ethyl ester | |
| ethyl 2-hydroxy-2,2-diphenylacetate |
Specifications
| 52182-15-7 | |
| White-Yellow | |
| C16H16O3 | |
| 5 g | |
| AIPVNQQMYPWQSX-UHFFFAOYSA-N | |
| ethyl 2-hydroxy-2,2-diphenylacetate | |
| 96261 | |
| ≥97.0% (GC) | |
| Ethyl Benzilate |
| 34°C | |
| 190°C | |
| MFCD00016850 | |
| ethyl benzilate, benzilic acid ethyl ester, ethyl diphenylglycolate, ethylbenzilate, ethyl benzillate, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, ethyl ester, benzilsaureethylester, acmc-209ky4, ethyl hydroxy diphenyl acetate #, diphenylglycolic acid ethyl ester | |
| CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 256.30 | |
| 256.30 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No