missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzilate 97.0+%, TCI America™

Supplier:  TCI America E074725G

 View more versions of this product

Catalog No. E074725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Benzilate
34°C
190°C
MFCD00016850
ethyl benzilate, benzilic acid ethyl ester, ethyl diphenylglycolate, ethylbenzilate, ethyl benzillate, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, ethyl ester, benzilsaureethylester, acmc-209ky4, ethyl hydroxy diphenyl acetate #, diphenylglycolic acid ethyl ester
CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
256.30
256.30
Crystalline Lumps
52182-15-7
White-Yellow
C16H16O3
25 g
AIPVNQQMYPWQSX-UHFFFAOYSA-N
ethyl 2-hydroxy-2,2-diphenylacetate
96261
≥97.0% (GC)

Chemical Identifiers

52182-15-7
256.30
AIPVNQQMYPWQSX-UHFFFAOYSA-N
96261
CCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
C16H16O3
MFCD00016850
ethyl benzilate, benzilic acid ethyl ester, ethyl diphenylglycolate, ethylbenzilate, ethyl benzillate, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, ethyl ester, benzilsaureethylester, acmc-209ky4, ethyl hydroxy diphenyl acetate #, diphenylglycolic acid ethyl ester
ethyl 2-hydroxy-2,2-diphenylacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions