missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, NF, 98-102%, Spectrum™ Chemical

ET105, 141-78-6, C4H8O2

$410.10 - $17157.77

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18602497
View Documents
Spectrum Chemical Mfg Cor
ET105500MLGL
500 mL Amber Glass Bottle
Each for $410.10
Only null left
 
18602498
View Documents
Spectrum Chemical Mfg Cor
ET105500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $1,995.83
Only null left
 
18602499
View Documents
Spectrum Chemical Mfg Cor
ET1054LTGL
4 L Amber Glass Bottle
Each for $1,408.65
Only null left
 
18602500
View Documents
Spectrum Chemical Mfg Cor
ET1054LTGLCS4
4 x 4 L Amber Glass Bottle
Case of 4 Each for $5,106.13
Only null left
 
18602501
View Documents
Spectrum Chemical Mfg Cor
ET10520LTMT
20 L Steel Can
Each for $4,115.33
Only null left
 
18602502
View Documents
Spectrum Chemical Mfg Cor
ET10555GLBL
55 gal. Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Ethyl Acetate, NF is used as a diluent and solvent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

141-78-6
88.11
ethyl acetate
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
CCOC(C)=O

Specifications

141-78-6
77°C
Amber Glass Bottle
UN1173
CCOC(C)=O
88.11
NF
100%
500 mL
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
98 to 102%
Ethyl Acetate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'