missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-nitroindole-2-carboxylate, 95%

Catalog No. AAH6665706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6665706 5 g
AAH6665703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6665706 Supplier Thermo Scientific Chemicals Supplier No. H6665706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6960-46-9 | C11H10N2O4 | 234.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6960-46-9
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.211
MDL Number MFCD00216475
InChI Key GTZAIVBXGPLYGD-UHFFFAOYSA-N
Synonym ethyl 7-nitroindole-2-carboxylate, 7-nitroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 7-nitro-, ethyl ester, ethyl-7-nitro-1h-indole-2-carboxylate, 2-carbethoxy-7-nitroindole, ethyl 7-nitro-2-indolecarboxylate, indole-2-carboxylic acid, 7-nitro-, ethyl ester, 1h-indole-2-carboxylicacid, 7-nitro-, ethyl ester
PubChem CID 250407
IUPAC Name ethyl 7-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C(=CC=C2)[N+](=O)[O-]

Specifications

Quantity 5 g
Beilstein 224930
Formula Weight 234.21
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 7-nitroindole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only