CAS RN 6960-46-9
IUPAC Name:
ethyl 7-nitro-1H-indole-2-carboxylate
Synonyms:
ethyl 7-nitro-1H-indole-2-carboxylate
Molecular Weight (g/mol):
234.21
Molecular Formula:
C11H10N2O4
InChi Key:
GTZAIVBXGPLYGD-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC2=CC=CC(=C2N1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Ethyl 7-nitroindole-2-carboxylate, 95%
Ethyl 7-nitroindole-2-carboxylate, 95%
CAS: 6960-46-9 Molecular Formula: C11H10N2O4 Molecular Weight (g/mol): 234.211 MDL Number: MFCD00216475 InChI Key: GTZAIVBXGPLYGD-UHFFFAOYSA-N Synonym: ethyl 7-nitroindole-2-carboxylate,7-nitroindole-2-carboxylic acid ethyl ester,1h-indole-2-carboxylic acid, 7-nitro-, ethyl ester,ethyl-7-nitro-1h-indole-2-carboxylate,2-carbethoxy-7-nitroindole,ethyl 7-nitro-2-indolecarboxylate,indole-2-carboxylic acid, 7-nitro-, ethyl ester,1h-indole-2-carboxylicacid, 7-nitro-, ethyl ester PubChem CID: 250407 IUPAC Name: ethyl 7-nitro-1H-indole-2-carboxylate SMILES: CCOC(=O)C1=CC2=C(N1)C(=CC=C2)[N+](=O)[O-]