missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH6356903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6356903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6356903 Supplier Thermo Scientific Chemicals Supplier No. H6356903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 16792-45-3
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.21
MDL Number MFCD04973967
InChI Key SOMZLSPJSSDTAP-UHFFFAOYSA-N
Synonym ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester
PubChem CID 616444
IUPAC Name ethyl 6-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 234.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 6-nitroindole-2-carboxylate
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only