CAS RN 16792-45-3
IUPAC Name:
ethyl 6-nitro-1H-indole-2-carboxylate
Synonyms:
ethyl 6-nitro-1H-indole-2-carboxylate
Molecular Weight (g/mol):
234.21
Molecular Formula:
C11H10N2O4
InChi Key:
SOMZLSPJSSDTAP-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals
CAS: 16792-45-3 Molecular Formula: C11H10N2O4 Molecular Weight (g/mol): 234.21 MDL Number: MFCD04973967 InChI Key: SOMZLSPJSSDTAP-UHFFFAOYSA-N Synonym: ethyl 6-nitroindole-2-carboxylate,6-nitro-1h-indole-2-carboxylic acid ethyl ester,6-nitro-2-carbethoxyindole,6-nitro-2-carboethoxyindole,ethyl 6-nitroindol-2-carboxylate,1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester,1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester PubChem CID: 616444 IUPAC Name: ethyl 6-nitro-1H-indole-2-carboxylate SMILES: CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O