missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Methylsalicylate 98.0+%, TCI America™

Catalog No. M12835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12835G 5 g
M128325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12835G Supplier TCI America Supplier No. M12835G

Chemical Identifiers

CAS 6555-40-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD01194299
InChI Key KWXBNUYCDMPLEQ-UHFFFAOYSA-N
Synonym Ethyl 2-Hydroxy-6-methylbenzoate, 2-Hydroxy-6-methylbenzoic Acid Ethyl Ester, 6-Methylsalicylic Acid Ethyl Ester
PubChem CID 584222
IUPAC Name ethyl 2-hydroxy-6-methylbenzoate
SMILES CCOC(=O)C1=C(C=CC=C1O)C

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 6-Methylsalicylate