accessibility menu, dialog, popup

UserName

CAS RN 6555-40-4

H3COOCH3HO

CAS RN 6555-40-4

IUPAC Name: ethyl 2-hydroxy-6-methylbenzoate
Synonyms: ethyl 2-hydroxy-6-methylbenzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: KWXBNUYCDMPLEQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results