missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-nitroindole-2-carboxylate, 97%

Catalog No. AAH6699703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6699703 1 g
AAH6699706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6699703 Supplier Thermo Scientific Chemicals Supplier No. H6699703

CAS: 16732-57-3 | C11H10N2O4 | 234.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16732-57-3
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.21
MDL Number MFCD00216477
InChI Key DVFJMQCNICEPAI-UHFFFAOYSA-N
Synonym ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7
PubChem CID 280312
IUPAC Name ethyl 5-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O

Specifications

Quantity 1 g
Beilstein 225290
Formula Weight 234.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 5-nitroindole-2-carboxylate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only