accessibility menu, dialog, popup

UserName

CAS RN 16732-57-3

IUPAC Name: ethyl 5-nitro-1H-indole-2-carboxylate
Synonyms: ethyl 5-nitro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 234.21
Molecular Formula: C11H10N2O4
InChi Key: DVFJMQCNICEPAI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)