missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-nitroindole-2-carboxylate, 95%

Catalog No. AC424660100
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424660100 10 g Glass bottle
AC424660025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC424660100 Supplier Thermo Scientific Chemicals Supplier No. 424660100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16732-57-3 | C11H10N2O4 | 234.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16732-57-3
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.21
MDL Number MFCD00216477
InChI Key DVFJMQCNICEPAI-UHFFFAOYSA-N
Synonym ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7
PubChem CID 280312
IUPAC Name ethyl 5-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O

Specifications

Melting Point 220°C to 225°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: negligible
Formula Weight 234.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ethyl 5-nitroindole-2-carboxylate
Show More Show Less

RUO – Research Use Only