missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-Chloroindole-2-carboxylate 98.0+%, TCI America™
Supplier: TCI America E069625G
Specifications
| Ethyl 5-Chloroindole-2-carboxylate | |
| 170°C | |
| C11H10ClNO2 | |
| 25 g | |
| LWKIFKYHCJAIAB-UHFFFAOYSA-N | |
| ethyl 5-chloro-1H-indole-2-carboxylate | |
| 78518 | |
| ≥98.0% (GC) |
| 4792-67-0 | |
| White-Yellow | |
| MFCD00005610 | |
| ethyl 5-chloroindole-2-carboxylate, ethyl 5-chloro-2-indolecarboxylate, 5-chloroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester, ethyl-5-chloroindole-2-carboxylate, 5-chloro-1h-indole-2-carboxylic acid ethyl ester, pubchem7173, 5-chloroindole-2-carboxylicacidethylester, acmc-1aoif, 5-chloro-2-indolecarboxylic acid ethyl ester | |
| CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl | |
| 223.656 | |
| 223.66 | |
| Crystalline Powder |
Chemical Identifiers
| 4792-67-0 | |
| 223.656 | |
| LWKIFKYHCJAIAB-UHFFFAOYSA-N | |
| 78518 | |
| CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl |
| C11H10ClNO2 | |
| MFCD00005610 | |
| ethyl 5-chloroindole-2-carboxylate, ethyl 5-chloro-2-indolecarboxylate, 5-chloroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester, ethyl-5-chloroindole-2-carboxylate, 5-chloro-1h-indole-2-carboxylic acid ethyl ester, pubchem7173, 5-chloroindole-2-carboxylicacidethylester, acmc-1aoif, 5-chloro-2-indolecarboxylic acid ethyl ester | |
| ethyl 5-chloro-1H-indole-2-carboxylate |
Safety and Handling
TSCA : No