missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Chloroindole-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7129108
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06965G 5 g
E069625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06965G Supplier TCI America Supplier No. E06965G

Chemical Identifiers

CAS 4792-67-0
Molecular Formula C11H10ClNO2
Molecular Weight (g/mol) 223.656
MDL Number MFCD00005610
InChI Key LWKIFKYHCJAIAB-UHFFFAOYSA-N
Synonym ethyl 5-chloroindole-2-carboxylate, ethyl 5-chloro-2-indolecarboxylate, 5-chloroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester, ethyl-5-chloroindole-2-carboxylate, 5-chloro-1h-indole-2-carboxylic acid ethyl ester, pubchem7173, 5-chloroindole-2-carboxylicacidethylester, acmc-1aoif, 5-chloro-2-indolecarboxylic acid ethyl ester
PubChem CID 78518
IUPAC Name ethyl 5-chloro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 223.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Chloroindole-2-carboxylate