missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(Trifluoromethyl)benzoate 98.0+%, TCI America™

Catalog No. p-7129436
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13305G 5 g
E133025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E13305G Supplier TCI America Supplier No. E13305G

Chemical Identifiers

CAS 583-02-8
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD00013563
InChI Key ZQDADDSPMCHZPX-UHFFFAOYSA-N
Synonym ethyl 4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid ethyl ester, 4-trifluoromethylbenzoic acid, ethyl ester, pubchem16913, acmc-1cs5c, ethyl-p-trifluoromethylbenzoate, ethyl-4-trifluoromethyl benzoate, 4-trifluoromethylbenzoic acid ethyl ester
PubChem CID 521827
IUPAC Name ethyl 4-(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 80°C
Quantity 5 g
Formula Weight 218.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-(Trifluoromethyl)benzoate